Learn More
Medchemexpress LLC Vardenafil N-oxide | 448184-48-3 | ≥95.0% | 504.60 | C23H32N6O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1337245MG
Encompass_Preferred
Vardenafil N-oxide is the N-oxide impurity of vardenafil provided as a research/reference standard for analytical and quality-control applications. It is intended for use in method development, impurity identification, and stability or metabolic studies where an authentic impurity standard is required.
  • Reference standard for analytical methods.
  • Suitable for impurity profiling and method validation.
  • Stable powder form with defined storage recommendations.
  • Molecular formula C23H32N6O5S; molecular weight 504.60.
  • Supplied in small quantities for analytical use.
Catalog No. 50-002-62655