Learn More
Medchemexpress LLC HY-10019 5mg Medchemexpress, Varenicline CAS:249296-44-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100195MG
Encompass_Preferred
Medchemexpress, HY-10019 5mg Varenicline CAS:249296-44-4 Varenicline (CP 526555) is a selective α4β2 nicotinic receptor (nAChR) partial agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1742