Learn More
Medchemexpress LLC Varenicline-d4 hydrochloride | 866823-63-4 | MFCD12546330 | 98.0% | 215.293646 | C13H9D4N3·xHCl | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HY10019AS12.5MG
Encompass_Preferred
Varenicline-d4 hydrochloride is a deuterium-labeled analytical standard supplied as a hydrochloride salt for use as an isotopic tracer and internal standard in quantitative assays. It is intended for research use in NMR, GC-MS, and LC-MS workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (98.0%).
  • Supplied as a small-quantity hydrochloride salt (2.5 mg).
  • Useful as a tracer in drug development and bioanalytical studies.
Catalog No. 50-002-15874