Learn More
Selleck Chemical LLC Varenicline Hydrochloride S5022-5mg
SDP
Supplier:  Selleck Chemical LLC S50225MG
Encompass_Preferred
Varenicline (CP 526555) Hydrochloride is a partial agonist of 4 2 nicotine acetylcholine receptor (nAChR) and 6 2 nicotine acetylcholine receptor (nAChR) with a Ki value of 0 12 nM and 0 14 nM for 6 2 nAChR and 4 2 nAChR respectively in the striatum of rats
Catalog No. 50-311-0514