Learn More
Selleck Chemical LLC Varespladib (LY315920) S1110-5mg
SDP
Supplier:  Selleck Chemical LLC S11105MG
Encompass_Preferred
Varespladib (LY315920) is a potent and selective human non-pancreatic secretory phospholipase A2 (hnsPLA) inhibitor with IC50 of 7 nM Phase 3
Catalog No. 50-310-8500