Learn More
Medchemexpress LLC Varespladib methyl | 172733-08-3 | 99.9% | 394.42 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1744810MM/1ML
Encompass_Preferred
Varespladib methyl (A-002; LY333013) is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920. It demonstrates high-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations against 28 medically important snake venoms from six continents.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2).
  • Orally-administered methyl ester pro-drug.
  • Metabolized to LY315920.
  • Exhibits high-level sPLA2 inhibition against 28 snake venoms.
  • Purity: 99.90%.
Catalog No. 50-003-86544