Learn More
Medchemexpress LLC Vari Fluor 647a carboxylic acid free acid | 98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD18231MG
Encompass_Preferred
Vari Fluor 647A Carboxylic acid free acid is a carboxylic acid derivative of Vari Fluor. This unactivated labeled fluorescent dye is designed for various labeling applications.
  • Used for protein labeling.
  • Used for antibody labeling.
  • Used for polysaccharide labeling.
  • Requires carboxylic acid activation for use.
Catalog No. 50-004-31146