Learn More
Instech Vascular Access Button™ for mouse, 1ch o-ring, silicone mesh with holes, 25ga, injector
SDP
Supplier:  Instech VAB62SMBS/25
Encompass
Silicone Mesh Vascular Access Button™ for Mice
For tethered and intermittent access
This VAB™ variant was developed for the surgeons at The Jackson Laboratory who needed a design that would work with their diverse strains of mice. The button provides a number of benefits over a simple exteriorized catheter:
  • Simplified access with a syringe for intermittent dosing or sampling, or a tether for continuous infusion
  • Improved patency
  • Ability to group house (with the protective cap, sold separately)
The button features a medical-grade silicone mesh disk, 14mm in diamter, with eight holes through which tissue will grow. The button stalk is our original o-ring based design which minimizes the exit site. It does not work with our tethers that attach with magnets; however, the silicone disk is also available on our standard magnetic mouse VAB.
The buttons have 25ga connectors to mate with 2Fr polyurethane mouse catheters.
Catalog No. NC2197412