Learn More
Medchemexpress LLC Vasculotide | 1359657-45-6 | 99.8% | 14000 (average) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP105801MG
Encompass_Preferred
Vasculotide is an angiopoietin-1 mimetic that acts as a Tie-2 activator, inducing Tie-2 phosphorylation. It has anti-inflammatory and anti-permeability effects. This product ameliorates endotoxin-induced endothelial barrier dysfunction, promotes angiogenesis in a mouse model of diabetic ulcer, protects mice from vascular leakage, reduces mortality in murine abdominal sepsis, decreases microvascular leakage, and improves microcirculatory perfusion in a rat model of hemorrhagic shock.
  • Acts as a Tie-2 activator
  • Induces Tie-2 phosphorylation
  • Exhibits anti-inflammatory effects
  • Exhibits anti-permeability effects
  • Ameliorates endothelial barrier dysfunction
  • Promotes angiogenesis
  • Protects from vascular leakage
  • Reduces mortality in abdominal sepsis
  • Decreases microvascular leakage
  • Improves microcirculatory perfusion
Catalog No. 50-004-24764