Learn More
Medchemexpress LLC Vasopressin Dimer (parallel) (TFA) | 99.55% | 2282.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP52135MG
Encompass_Preferred
Vasopressin Dimer (parallel) TFA is a parallel dimer of Vasopressin. It can activate four G protein-coupled receptors: V1aR, V1bR, V2R, and OTR.
  • Purity of 99.55%
  • Solid appearance
  • White to off-white color
  • Suitable for metabolic or endocrine disease research
  • Activates V2 Receptor
Catalog No. 50-003-99184