Learn More
CPC Scientific H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Arg-Gly-NH2
(Disulfide bridge) 25MG
SDP
Supplier:  CPC Scientific OXYT005B
Encompass
Vasotocin also named argiprestocin is the vasopressin analog produced in birds, amphibians and fish. The peptide shows antidiuretic as well as reproductive activities.SEQUENCE: H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Arg-Gly-NH2(Disulfide bond)ONE-LETTER SEQUENCE: CYIQNCPRG-NH2 (Disulfide bridge)MOLECULAR FORMULA: C43H67N15O12S2MOLECULAR WEIGHT: 1050.22STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [74927-14-3]SYNONYMS: Argiprestocin, (Arg8)-VasotocinRESEARCH AREA: HormonalREFERENCES:F.L.Moore and C.A.Lowry, Comp. Biochem. Physiol. C, Pharmacol. Toxicol. Endocrinol., 119, 251 (1998)A.Jurkevich and R.Grossmann, Domest. Anim. Endcrinol., 25, 93 (2003)F.Toyoda et al., Horm. Behav., 44, 346 (2003)R.J.Balment et al., Gen. Comp. Endocrinol., 147, 9 (2006)SKU(s): OXYT-005A, OXYT-005B
Catalog No. 50-211-0053