Learn More
Medchemexpress LLC Vatinoxan (hydrochloride) | 130466-38-5 | MFCD08056257 | 99.8% | 10 MG

Supplier: Medchemexpress LLC HY19057A10MG
Vatinoxan hydrochloride is the hydrochloride salt of vatinoxan, a peripherally selective α2-adrenergic receptor antagonist supplied as a high-purity solid for research use in pharmacology and in vivo studies.
- Peripherally selective α2-adrenergic receptor antagonist for pharmacological studies.
- High purity (≈99.8%) suitable for analytical and in vivo use.
- Hydrochloride salt supplied as a solid with good stability under recommended storage conditions.
- Soluble in DMSO (~10 mg/mL); ultrasonic agitation may be required.
- Recommended storage: refrigerated under inert atmosphere; solution stability at -80°C (6 months) or -20°C (1 month).
- Available in multiple small research quantities for dosing and formulation studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.