Learn More
Medchemexpress LLC VAV1 degrader-2 | 3050683-29-6 | 98.2% | 393.87 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1686245MG
Encompass_Preferred
VAV1 degrader-2 (Example 176) is a molecular glue degrader that targets VAV 1 with a DC50 of 4.41 nM. It can be used in the study of inflammatory and autoimmune diseases.
  • Molecular glue degrader
  • Targets VAV 1
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
  • Useful in the study of inflammatory and autoimmune diseases
Catalog No. 50-003-93765