Learn More
Medchemexpress LLC VB124 (MCT4 inhibitor) | 2230186-18-0 | MFCD34602725 | 98.9% | C23H23ClN2O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13966510MG
Encompass_Preferred
VB124 is an orally active, potent, and selective monocarboxylate transporter 4 (MCT4) inhibitor used in research to block lactate import and export. It exhibits low-nanomolar cellular activity (IC50 = 8.6 nM for import, 19 nM for export in MDA-MB-231 cells) and is applied in studies of cancer metabolism, cardiac hypertrophy, and heart failure.
  • Selective MCT4 inhibition.
  • Low-nanomolar potency in cellular assays.
  • Redirects glycolytic carbon flux toward mitochondria.
  • Available as solid and DMSO solution formulations.
  • Recommended storage for powder and solvent formulations.
  • High reported purity (~98.9%).
Catalog No. 50-002-61696