Learn More
Medchemexpress LLC VBIT-4 | 2086257-77-2 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129122100MG
Encompass_Preferred
VBIT-4 is an inhibitor of voltage-dependent anion channel 1 (VDAC1) oligomerization with a binding affinity (Kd) of 17 μM. It acts as an apoptosis inhibitor and can be used for therapeutic purposes in apoptosis-associated disorders such as neurodegenerative and cardiovascular diseases. It targets the mitochondrial protein VDAC1, inhibiting apoptosis and protecting against mitochondrial dysfunction.
  • Inhibitor of VDAC1 oligomerization
  • Apoptosis inhibitor
  • Protects against mitochondrial dysfunction
  • Can be used for therapeutic purposes in apoptosis-associated disorders
Catalog No. 50-003-39372