Learn More
TARGETMOL CHEMICALS INC VBY-825 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T132881MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. VBY-825 is a reversible inhibitor of cathepsin with high potency against cathepsins B, L, S and V. Purity 98.99%
Catalog No. 50-287-6134