Learn More
Medchemexpress LLC VCAM-1/CD106 Human 5ug
SDP
Supplier:  Medchemexpress LLC HYP734685UG
Encompass_Preferred
VCAM-1/CD106 proteins expressed on endothelial cells critically regulate leukocyte adhesion through integrin interactions initiating calcium channel activation and RAC1-mediated transendothelial migration during inflammation VCAM-1/CD106 Protein Human (HEK293 Fc) is the recombinant human-derived VCAM-1/CD106 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-80316