Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Lithium Heparin Additive, 4 mL, Hemogard™ Closure Plastic Tube, 100/Box
Supplier:  Med Vet International 367884
Encompass_Preferred
Features Lithium Heparin, spray dried, 75 USP units
Catalog No. 50-304-3232