Learn More
Santa Cruz Biotechnology VDAC1 Antibody (B-6), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC390996
Encompass
VDAC1 Antibody (B-6) is a high quality monoclonal VDAC1 antibody (also designated Voltage dependent anion channel 1 antibody, PORIN antibody, or VDAC-1 antibody) suitable for the detection of the VDAC1 protein of mouse, rat and human origin. VDAC1 Antibody (B-6) is available as both the non-conjugated anti-VDAC1 antibody form, as well as multiple conjugated forms of anti-VDAC1 antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1395951