Learn More
Apexbio Technology LLC VE-822 1232416-25-9 50mg
SDP
Supplier:  Apexbio Technology LLC B138350
Encompass_Preferred
VE-822 (CAS 1232416-25-9) is a potent small molecule inhibitor of ATR (ATM-Rad3-related kinase) exhibiting an IC50 value of 0 019 M By specifically targeting ATR VE-822 interferes with DNA damage response mechanisms triggered by replication stress and DNA double-strand breaks processes frequently activated following chemotherapy or radiotherapy (XRT) Preclinical studies demonstrate that VE-822 disrupts cell cycle checkpoint signaling and homologous recombination enhancing persistent DNA damage in cancer cells In pancreatic tumor xenograft models VE-822 selectively sensitizes tumors rather than normal tissues to radiation and gemcitabine treatments highlighting its potential utility in oncology research aimed at exploiting DNA repair pathway vulnerabilities
Catalog No. 50-000-63360