Learn More
Santa Cruz Biotechnology Anti-VE-cadherin Antibody (F-8), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC9989
Encompass
Anti-VE-cadherin Antibody (F-8) is recommended for detection of VE-cadherin of mouse, rat and human origin by WB, IP, IF, FCM and ELISA; also reactive with additional species, including porcine.
Catalog No. NC1103782