Learn More
Medchemexpress LLC VE-Cadherin Human 50ug
SDP
Supplier:  Medchemexpress LLC HYP73560A50UG
Encompass_Preferred
The FGFR-4 protein binds to its ligands and activates signaling pathways involved in cell growth differentiation and survival It plays a role in various developmental processes including skeletal development liver metabolism and cancer progression In the absence of ligand binding FGFR-4 remains inactive and does not initiate downstream signaling cascades VE-Cadherin Protein Human (HEK293 C-His-Fc) is the recombinant human-derived VE-Cadherin protein expressed by HEK293 with C-hFc C-6 His labeled tag
Catalog No. 50-001-80257