Learn More
Medchemexpress LLC VE-Cadherin Mouse 5ug
SDP
Supplier:  Medchemexpress LLC HYP735595UG
Encompass_Preferred
VE-cadherin is a calcium-dependent glycoprotein critical for mediating cell adhesion and coordinating morphogenetic events during development Lack of VE-cadherin in mice leads to uterine death due to increased endothelial cell apoptosis leading to vascular insufficiency VE-Cadherin Protein Mouse (HEK293 His) is the recombinant mouse-derived VE-Cadherin protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-80081