Learn More
Medchemexpress LLC Vecabrutinib | 1510829-06-7 | 99.76% | 529.92 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10907850MG
Encompass_Preferred
Vecabrutinib (SNS-062) is a potent, noncovalent BTK and ITK inhibitor. It demonstrates high specificity and activity, making it suitable for research applications.
  • Potent BTK and ITK inhibitor
  • Noncovalent binding
  • Specific Kd values for BTK (0.3 nM) and ITK (2.2 nM)
  • IC50 of 24 nM for ITK
Catalog No. 50-003-44429