Learn More
Vector Laboratories VECTASHIELD HardSet™ Antifade Mounting Medium With DAPI 10 ml
SDP
Supplier:  Vector Laboratories H150010
Encompass
VECTASHIELDHardSet™ Mounting Medium prevents rapid photobleaching of fluorescent proteins and dyes. After approximately 15 minutes at room temperature, the mounting medium will harden, the coverslip will become immobilized, and optimal antifade ability and refractive index of 1.46 will be achieved. Mounted slides should be stored at 4 degrees C, protected from light. VECTASHIELD HardSet Mounting Medium is compatible with a wide array of fluorochromes, enzymatic substrates, and fluorescent proteins. The refractive index is 1.45. DAPI is a counterstain that binds to nucleii and causes them to fluoresce with a blue color. This is useful for Fluorescence In Situ Hybridization (FISH) assays and other techniques where a general orientation to the location of cells and nucleii is helpful. An attractive alternative to VECTASHIELD HardSet with DAPI is VECTASHIELD Vibrance™ with DAPI (NC1601055).
Catalog No. NC9029229
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.