Learn More
TARGETMOL CHEMICALS INC Vecuronium bromide 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1594500MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vecuronium bromide (ORG NC 45) is a synthetic, intermediate-acting, mono-quaternary steroid that functions as a non-depolarizing neuromuscular blocking agent with muscle relaxant properties. Purity 99.92%
Catalog No. 50-287-7524