Learn More
Medchemexpress LLC Vecuronium bromide | 50700-72-6 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0118A10MM/1ML
Encompass_Preferred
Vecuronium bromide is a non-depolarizing neuromuscular blocking agent. It also functions as a nicotinic acetylcholine receptor (nAChR) inhibitor and a muscle relaxant. This compound can be utilized for pre-surgical anesthesia.
  • Acts as a nicotinic acetylcholine receptor (nAChR) inhibitor
  • Functions as a muscle relaxant
  • Can be used for pre-surgical anesthesia
  • Inhibits human MATE1-mediated ASP+ uptake
  • Inhibits human MATE2K-mediated ASP+ uptake
  • Inhibits human OCT2-mediated ASP+ uptake
  • Inhibits cancer cell proliferation and migration
Catalog No. 50-004-50845