Learn More
Selleck Chemical LLC Vecuronium Bromide 100mg 50700-72-6 ORG NC45
SDP
Supplier:  Selleck Chemical LLC S1405100MG
Encompass_Preferred
Vecuronium Bromide (ORG NC45) is a non-depolarizing neuromuscular blocking agent, used for skeletal muscle relaxation during surgery. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-818-5