Learn More
Cayman Chemical VecuronIumbromIde 10mg
SDP
Supplier:  Cayman Chemical 1560310 MG
Encompass_Preferred
A non-depolarizing muscle relaxant that competitively blocks cholinergic receptors at the motor end plate of the neuromuscular junction, inducing temporary paralysis
Catalog No. 50-297-2633