Learn More
TARGETMOL CHEMICALS INC Vedaprofen 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3668310MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vedaprofen (PM 150) inhibits COX-1 and reduces prostaglandin H2 synthesis with anti-inflammatory activities. Vedaprofen is an inhibitor of Escherichia coli sliding clamp with the IC50 of 222 uM and Ki of 131 uM. Purity 98.12%
Catalog No. 50-287-8773