Learn More
Medchemexpress LLC Vedaprofen (standard) | 71109-09-6 | MFCD00866019 | 282.38 g/mol | C19H22O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1188275MG
Encompass_Preferred
Vedaprofen is a COX-1 selective nonsteroidal anti-inflammatory drug supplied as a research standard for analytical and preclinical studies. The compound has molecular formula C19H22O2 and molecular weight 282.38 g/mol, and is offered in small, accurately measured pack sizes for assay and reference use.
  • COX-1 selective nonsteroidal anti-inflammatory agent.
  • Suitable as a reference standard for analytical and pharmacological studies.
  • Molecular formula C19H22O2; molecular weight 282.38 g/mol.
  • Available in small pack sizes for precise dosing and assay preparation.
  • Includes datasheet and safety data sheet documentation for handling and storage.
Catalog No. 50-002-29008