Learn More
MyBioSource VEGF-C ELISA Kit
SDP
Supplier:  MyBioSource MBS738525
Encompass_Preferred
The MyBioSource Human VEGF-C ELISA Kit quantifies VEGF-C in various biological fluids using a specific antibody-coated 96-well plate. Samples and standards are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and TMB substrate for colorimetric measurement at 450 nm.
Catalog No. NC2739385