Learn More
Medchemexpress LLC VEGF-C Human HEK29 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7786420UG
Encompass_Preferred
VEGF-C protein is an important member of the PDGF/VEGF growth factor family and plays a role in important cell signaling pathways As part of this family VEGF-C may share features with related proteins that promote cell growth angiogenesis and lymphangiogenesis VEGF-C Protein Human (HEK293 His-Avi) is the recombinant human-derived VEGF-C protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87518