Learn More
Medchemexpress LLC VEGF-D Rat HEK293 2ug
SDP
Supplier:  Medchemexpress LLC HYP734752UG
Encompass_Preferred
VEGF-D a growth factor stimulates proliferation migration and vessel permeability It is crucial for vascular development and maintenance of lymphatic endothelium VEGF-D binds and activates VEGFR-3 initiating essential signaling pathways Structurally it forms a non-covalent homodimer playing an intricate role in vascular processes VEGF-D Protein Rat (HEK293 Fc) is the recombinant rat-derived VEGF-D protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80679