Learn More
Medchemexpress LLC VEGF Receptor 2 Antibody | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8164310UL
Encompass_Preferred
VEGF Receptor 2 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody that targets VEGF Receptor 2. It is also known by several synonyms including KDR, FLK1, VEGFR2, and CD309, indicating its role in vascular endothelial growth factor signaling.
  • Applications include Western Blot, Immunohistochemistry (paraffin-embedded and frozen sections), Immunocytochemistry/Immunofluorescence, and ELISA
  • Reactive with human and mouse samples
Catalog No. 50-004-41360