Learn More
Medchemexpress LLC HY-P7110A 50ug Medchemexpress, VEGF165 (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP7110A50UG
Encompass_Preferred
Medchemexpress, HY-P7110A 50ug VEGF165 (HEK293-expressed), Human CAS: Recombinant Human Vascular Endothelial Growth Factor 165 (HEK293-expressed) is a Vascular Endothelial Growth Factor A (VEGF-A) isoform. VEGF is a heparin-binding growth factor specific for vascular endothelial cells that is able to induce angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4340