Learn More
Medchemexpress LLC VEGFR-1 Human 328a 100ug
SDP
Supplier:  Medchemexpress LLC HYP73065100UG
Encompass_Preferred
The VEGFR-1 protein is a tyrosine protein kinase that acts as a cell surface receptor for VEGFA VEGFB and PGF It plays a crucial role in embryonic vasculature development regulation of angiogenesis cell survival migration macrophage function chemotaxis and cancer cell invasion VEGFR-1 Protein Human (328a a HEK293 His) is the recombinant human-derived VEGFR-1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80797