Learn More
ACROBiosystems Recombinant Protein;Human VEGF R3 / FLT4 Protein, Fc Tag;HEK293;1MG;FL4,Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL,VEGF R3
Supplier:  ACROBiosystems FL4H52511MG
Encompass_Preferred
Recombinant Protein;Human VEGF R3 / FLT4 Protein, Fc Tag;1MG;Human VEGF R3, Fc Tag (FL4-H5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # NP_002011.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 111.2 kDa. As a result of glycosylation, the protein migrates as 140-160 kDa under reducing (R) condition, and 280-320 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5μg/mL (100 μL/well) can bind Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) at 5μg/mL (100 μL/well) can bind Human VEGF-D, His Tag (Cat. No. VED-H5228) with a linear range of 0.08-0.6 μg/mL (Routinely tested).
Catalog No. 50-165-8460