Learn More
Medchemexpress LLC HY-10129 10mg Medchemexpress, Veliparib CAS:912444-00-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012910MG
Encompass_Preferred
Medchemexpress, HY-10129 10mg Veliparib CAS:912444-00-9 Veliparib (ABT-888) is a potent PARP inhibitor, inhibiting PARP1 and PARP2 with Kis of 5.2 and 2.9 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8657