Learn More
Medchemexpress LLC Veliparib (ABT-888) | 912444-00-9 | 99.8% | 244.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY10129
Encompass_Preferred
Veliparib (ABT-888) is a potent PARP inhibitor that targets PARP1 and PARP2 with high specificity. It demonstrates good oral bioavailability and the ability to cross the blood-brain barrier in various tumor models, making it suitable for a range of research applications.
  • Potent PARP inhibitor
  • Inhibits PARP1 and PARP2 with Ki values of 5.2 nM and 2.9 nM, respectively
  • Good oral bioavailability
  • Can cross the blood-brain barrier
  • Targets autophagy
Catalog No. 50-003-64755