Learn More
Selleck Chemical LLC Veliparib (ABT-888) 10mM/1mL 912444-00-9 NSC 737664
SDP
Supplier:  Selleck Chemical LLC S100410MM/1ML
Encompass_Preferred
Veliparib (ABT-888, NSC 737664) is a potent inhibitor of PARP1 and PARP2 with Ki of 5.2 nM and 2.9 nM in cell-free assays, respectively. It is inactive to SIRT2. Veliparib increases autophagy and apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0968990