Learn More
Cole-Parmer Velp F206A0179 Compact Magnetic Stirrer without Motor; 120/240V 50/60Hz
Supplier:  Cole-Parmer 8657900
Encompass
Stirring action is produced by creating a rotating magnetic field with a series of electric coils at varied current. Unit has no moving parts and requires no maintenance. "Soft start" allows for optimal progression to desired stirring speed. Ultrathin stirrer is ideal for use in hoods or cabinets and can also be used in incubators.
Catalog No. NC1703073