Learn More
TARGETMOL CHEMICALS INC Venlafaxine 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2245350MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Venlafaxine (Wy 45030), an arylalkanolamine serotonin-norepinephrine reuptake inhibitor (SNRI), is used to treat major generalised anxiety disorder (GAD), depressive disorder (MDD), panic disorder and social phobia. Purity 99.2%
Catalog No. 50-287-7732