Learn More
Medchemexpress LLC Venlafaxine-d6 | 1020720-02-8 | 98.55% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0196S5MG
Encompass_Preferred
Venlafaxine-d6 is the deuterium labeled Venlafaxine. Venlafaxine (Wy 45030) is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor and an antidepressant. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules
Catalog No. 50-004-07420