Learn More
Medchemexpress LLC Venlafaxine-d6 hydrochloride | 1062606-12-5 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0196AS15MG
Encompass_Preferred
Venlafaxine-d6 hydrochloride is the deuterium labeled Venlafaxine hydrochloride. It is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor and an antidepressant. It is intended for research purposes only.
  • Deuterium labeled compound
  • Orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor
  • Antidepressant
  • Used for research purposes only
  • Suitable as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05568