Learn More
Medchemexpress LLC Venlafaxine (hydrochloride) | 99300-78-4 | 99.76% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0196A10MM/1ML
Encompass_Preferred
Venlafaxine hydrochloride is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor and an antidepressant.
  • Potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor
  • Antidepressant properties
  • 99.76% purity
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
  • Orally active
Catalog No. 50-004-07033