Learn More
Medchemexpress LLC Venlafaxine hydrochloride | 99300-78-4 | 99.8% | 25 MG
Supplier: Medchemexpress LLC 25MGHYB0196AR
Venlafaxine hydrochloride is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor. It is an antidepressant used for research purposes only.
- Potent serotonin/norepinephrine reuptake dual inhibitor
- Antidepressant for research purposes
- Inhibits [3H]-5-HT reuptake in human SERT transfected cells (IC50: 0.2 μM)
- Inhibits [3H]-NE reuptake in human NET transfected cells (IC50: 2.55 μM)
- Inhibits human DAT expressed in HEK293 cells by neurotransmitter reuptake assay (IC50: 15700 nM)
- Dose-dependently blocks depletion of norepinephrine levels in rat hypothalamus