Learn More
Medchemexpress LLC Venlafaxine (hydrochloride) | 99300-78-4 | MFCD03658865 | 99.99% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0196AR5MG
Encompass_Preferred
Venlafaxine hydrochloride (Standard) is an analytical standard of Venlafaxine hydrochloride, intended for research and analytical applications. It is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor with antidepressant properties.
  • Analytical standard for research and analytical applications
  • Orally active
  • Potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor
  • Antidepressant properties
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used in HPLC, GC, and MS applications
Catalog No. 50-004-07384