Learn More
Roche Diagnostics Ventana Antibody Diluent with Casein
Supplier:  Roche Diagnostics 06440002001
Encompass
VENTANA Antibody Diluent with Casein is a buffered, proteinaceous solution containing salt and immunoglobulins used to dilute rabbit and mouse primary antibodies for use on the BenchMark, BenchMark GX, BenchMark XT, and BenchMark ULTRA automated staining platforms. This product is intended for in vitro diagnostic use.
Catalog No. NC1706198