Learn More
Medchemexpress LLC HY-14275 50mg , Verapamil CAS:52-53-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1427550MG
Encompass_Preferred
HY-14275 50mg Verapamil CAS: 52-53-9 Verapamil ((±)-Verapamil) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor. Verapamil also inhibits CYP3A4. Verapamil has the potential for high blood pressure, heart arrhythmias and angina research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9516